Synthware™ Distillation Connecting Adapters, Angled at 75°
For use as a component in atmospheric or vacuum distillations
Supplier:  Synthware™ A354914
 View more versions of this product
Catalog No. 31500662
Explore available promotions
Description
Description
  • Standard taper outer joint at the top
  • Inner joints on bottom and side arm are at a 75° angle to one another
Specifications
Specifications
Borosilicate Glass (3.3)
Distillation Connecting Adapter
Joints: 14/20 (Top), 24/40 (Bottom), 19/22 (Side)