missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthware™ Distillation Receiver, Dean Stark, Vacuum Jacketed
Dean Stark distillation receiver for easy separation of water from reaction or distillation system
$368.79 - $459.00
Catalog Number | Mfr. No. | Height (Metric) | Width (Metric) | Capacity | Standard Taper Joint | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (Metric) | Width (Metric) | Capacity | Standard Taper Joint | Price | Quantity | ||
31501070
|
Synthware™
D441410 |
190mm | 65mm | 10 mL | 14/20 |
Each for $384.79
|
|
||
31501071
|
Synthware™
D442410 |
275mm | 95mm | 10 mL | 24/40 |
Each for $427.56
|
|
||
31501072
|
Synthware™
D442420 |
275mm | 95mm | 20 mL | 24/40 |
Each for $459.00
|
|
||
31501069
|
Synthware™
D441405 |
205mm | 65mm | 5 mL | 14/20 |
Each for $368.79
|
|
||
Description
- Graduated
- Uptake tube vacuum jacketed to improve distillation performance
- Jacket eliminates possibility of condensation or recrystallization of distillate in sidearm
- With upper outer and lower inner standard taper joints, both of same size
- PTFE stopcock with 2 mm bore
Specifications
Distillation Receiver |