Synthware™ Erlenmeyer Flasks with Outer Joint
With flat bottom and narrow neck
Supplier:  Synthware™ F661425
 View more versions of this product
Catalog No. 31501318
Explore available promotions
Description
Description
  • With standard taper outer joint
  • Hand blown from tubing to ensure heavy and uniform wall
Specifications
Specifications
Erlenmeyer Flask
25 mL
14/20