Synthware™ Erlenmeyer Flasks with Outer Joint
With flat bottom and narrow neck
Supplier:  Synthware™ F661450
 View more versions of this product
Catalog No. 31501319
Explore available promotions
Description
Description
  • With standard taper outer joint
  • Hand blown from tubing to ensure heavy and uniform wall
Specifications
Specifications
Erlenmeyer Flask
50 mL
14/20