Synthware™ Erlenmeyer Flasks with Outer Joint
With flat bottom and narrow neck
Supplier: Synthware™ F668000
Description
- With standard taper outer joint
- Hand blown from tubing to ensure heavy and uniform wall
Specifications
Erlenmeyer Flask | |
1000 mL | |
29/42 |