Synthware™ Erlenmeyer Flasks, Threaded
For microscale applications
Supplier:  Synthware™ F501010
 View more versions of this product
Catalog No. 31502128
Explore available promotions
Description
Description
  • With 14/20 threaded joint
  • Supplied with one holed Nylon compression cap, one Viton O-ring and one PTFE-faced silicone rubber septum
Specifications
Specifications
Erlenmeyer Flask
10 mL
14/10
Product Suggestions
Product Suggestions