Synthware™ Extraction Thimble, Glass
For use with Soxhlet extractor
$81.88 - $87.53
Specifications
Description For use with Soxhlet extractor
Material Borosilicate Glass
Wall Thickness 1.5 mm
View More Specs
Products 6
Catalog Number Mfr. No. For Use With (Equipment) Diameter (Metric) Outer Height (Metric) Porosity Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Diameter (Metric) Outer Height (Metric) Porosity Price Quantity  
31501081
View Documents Promotion Details
Synthware™
E113590C
Fits Extractor Size: Medium 35 mm 90 mm Coarse
Each for $84.59
31501080
View Documents Promotion Details
Synthware™
E112585X
Fits Extractor Size: Small 25 mm 85 mm Extra Coarse
Each for $81.88
31501079
View Documents Promotion Details
Synthware™
E112585C
Fits Extractor Size: Small 25 mm 85 mm Course
Each for $81.88
31501082
View Documents Promotion Details
Synthware™
E113590X
Fits Extractor Size: Medium 35 mm 90 mm Extra Coarse
Each for $84.59
31501083
View Documents Promotion Details
Synthware™
E114513C
Fits Extractor Size: Large 45 mm 130 mm Coarse
Each for $87.53
31501084
View Documents Promotion Details
Synthware™
E114513X
Fits Extractor Size: Large 45 mm 130 mm Extra Coarse
Each for $87.53
Description
Description
  • All-glass extraction thimble permits viewing extraction process and subsequent weighing
  • Available in coarse and extra coarse porosities
Specifications
Specifications
For use with Soxhlet extractor
Borosilicate Glass
1.5 mm
SDS
missing translation for 'documents'
missing translation for 'documents'