Synthware™ Flat Bottom Flask, Single Neck
All sizes are hand blown from tubing to ensure a uniform wall thickness
Supplier:  Synthware™ F124100
 View more versions of this product
Catalog No. 31501132
Explore available promotions
Description
Description
  • Flat bottom, single short neck flask with standard taper outer joint
  • Heavy wall design
Specifications
Specifications
Joint: 24/40; Single neck
100 mL