Synthware™ Flat Bottom Flask, Single Neck
All sizes are hand blown from tubing to ensure a uniform wall thickness
Supplier: Synthware™ F124250
Description
- Flat bottom, single short neck flask with standard taper outer joint
- Heavy wall design
Specifications
Joint: 24/40; Single neck | |
250 mL |