missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthware™ Gas Dispersion Tubes, Fritted, 90° Angle

For use with filtration techniques

Supplier:  Synthware™ F284060C

 View more versions of this product

Catalog No. 31501554



Description

Description

  • 90° angle permits bubbles to be spread over a large area
  • With stem O.D. of 8 mm and Length: 150 mm
Specifications

Specifications

Porosity: Coarse
Coarse
40 mm
SDS
Documents

Documents

Promotions

Promotions