Synthware™ Mineral Oil Bubbler
Mineral Oil Bubbler used to monitor gas flow to reactions while providing a vent to the atmosphere.
Supplier:  Synthware™ B257040
 View more versions of this product
Catalog No. 31500701
Explore available promotions
Description
Description
  • With 10 mm O.D. hose connection.
Specifications
Specifications
Borosilicate Glass (3.3)
1 in.
7.3 in.
3.1 in.
Oil Bubbler
26 mm
185 mm
80 mm
Product Suggestions
Product Suggestions