Synthware™ Rotary Evaporator Anti-Splash Adapters, Modified
With two 4 mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask
Supplier: Synthware™ A602914
Description
Introduction: Anti-splash adapter is placed between the vapor tube and the evaporation flask on a rotary evaporator to prevent contents of the flask to be drawn into the condenser in the event of bumpingSpecifications
100 mL | |
Organic Synthesis | |
Borosilicate Glass (3.3) |
Anti-splash Adapter | |
Upper: 29/42, Lower: 14/20 |