Synthware™ Single Neck Pear Shaped Flasks
With standard taper outer joint
Supplier:  Synthware™ F321405L
 View more versions of this product
Catalog No. 31501187
Explore available promotions
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
Single neck pear shaped flask
5 mL
14/20