Synthware™ Single Neck Pear Shaped Flasks
With standard taper outer joint
Supplier: Synthware™ F322450L
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Single neck pear shaped flask | |
50 mL | |
24/40 |