missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthware™ Storage Flasks with Side Arm
Supplied with high vacuum valve with PTFE O-ring at tip
$132.18 - $234.97
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass (3.3) |
For Use With (Application) | Organic Synthesis |
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Description | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Description | Price | Quantity | |||
31501388
|
Synthware™
F461010 |
10 mL | 14/20 | Valve Size: 0-4mm |
Each for $132.18
|
|
|||
31501389
|
Synthware™
F461025 |
25 mL | 14/20 | Valve Size: 0-4mm |
Each for $138.94
|
|
|||
31501390
|
Synthware™
F461050 |
50 mL | 14/20 | Valve Size: 0-4mm |
Each for $175.71
|
|
|||
31501391
|
Synthware™
F461100 |
100 mL | 14/20 | Valve Size: 0-4mm |
Each for $175.71
|
|
|||
31501392
|
Synthware™
F464100 |
100 mL | 24/40 | Valve Size: 0-8mm |
Each for $179.62
|
|
|||
31501393
|
Synthware™
F461250 |
250 mL | 14/20 | Valve Size: 0-4mm |
Each for $194.97
|
|
|||
31501394
|
Synthware™
F464250 |
250 mL | 24/40 | Valve Size: 0-4mm |
Each for $199.06
|
|
|||
31501396
|
Synthware™
F464500 |
500 mL | 24/40 | Valve Size: 0-4mm |
Each for $200.32
|
|
|||
31501397
|
Synthware™
F464508 |
500 mL | 24/40 | Valve Size: 0-8mm |
Each for $218.03
|
|
|||
31501395
|
Synthware™
F461500 |
500 mL | 14/20 | Valve Size: 0-4mm |
Each for $187.88
|
|
|||
31501398
|
Synthware™
F464008 |
1000 mL | 24/40 | Valve Size: 0-8mm |
Each for $234.97
|
|
|||
Description
SYNTHWARE storage flask with standard taper outer joint and high vacuum valve. Valve has a PTFE O-ring at the tip. Valve is used to control liquid or gas flow into the flask.Specifications
Clear | |
Borosilicate Glass (3.3) | |
Organic Synthesis |