missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthware™ Powder Funnels, 60° Offset

Short neck with standard taper outer joint on both center and angled side neck

Supplier:  Synthware™ F392475

 View more versions of this product

Catalog No. 31501479


Explore available promotions

Description

Description

  • All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications

Specifications

75mm
Glass
Powder funnel, 60° offset
Videos
SDS
Documents

Documents

Promotions

Promotions