Synthware™ Vacuum or Argon Adapters
With hose connection on the side and male joint
Supplier: Synthware™ A292440
Description
Synthware vacuum or inert gas adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection is at a 90 degree angle to the standard taper inner joint.Specifications
Organic Synthesis | |
Vacuum or Argon Adapter | |
24/40 |
Borosilicate Glass (3.3) | |
1 |