missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ SYTOX™ Blue Dead Cell Stain, for flow cytometry DFS Item

SYTOX™ Blue dead cell stain is a simple and quantitative single-step dead-cell indicator for use with violet laser equipped flow cytometers.

Supplier:  Molecular Probes™ S34857

Catalog No. 501137613


Explore available promotions

Description

Description

  • A simple and quantitative single-step dead-cell indicator for use with violet laser equipped flow cytometers
  • SYTOX™ Blue dead cell stain is a high-affinity nucleic acid stain that easily penetrates cells with compromised plasma membranes but will not cross uncompromised cell membranes
  • After brief incubation with SYTOX™ Blue stain, the nucleic acids of dead cells fluoresce bright blue when excited with 405 nm violet laser light
  • The violet-excited fluorescence emission permits clear discrimination from probes excited by most other laser lines, facilitating the development of multicolor assays with minimal spectral overlap between signals
  • Supplied as a 1 mM solution in dimethylsulfoxide (DMSO)
Specifications

Specifications

1 mM
Fluorescence
Flow Cytometer
SYTOX™ Blue
Solution
SYTOX™
Room Temperature
Contains 1 vial of SYTOX™ blue dead cell stain (1 mM solution in DMSO).

Store in freezer (-5 to -30°C), desiccated and protect from light.
Viability Assay
Dead Cell Stain
444/480
Tube(s)
1 mL
Nucleus, Nucleic Acids
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.