Lovibond™ T-CAL™ Turbidity Standard, <0.1 NTU
T-CAL™ Turbidity Standards are stabilized Formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. Eliminate the opportunity for inaccurate turbidimeter calibrations resulting from the improper dilution or handling of Formazin stock solution.
Supplier:  Lovibond™ 48012099
 View more versions of this product
Catalog No. 15-398-360
Explore available promotions
Description
Description
  • Save time and eliminate errors - no need for dilution
  • EPA approved primary turbidity standard
  • Long shelf life
  • All concentrations of T-CAL™ Turbidity Standards are verified to meet tight quality control specifications, are EPA and ISO compliant for reporting purposes and guaranteed to save you time. As a primary standard, T-CAL™ Turbidity Standards can be used to calibrate or verify any manufacturer's turbidimeter.
Specifications
Specifications
Turbidimeter
Liquid
<0.1 NTU
1000 mL/1 L
Amber Plastic Bottles
Primary
Calibration or Verification of a Turbidimeter
EN ISO 7027, USEPA 180.1 and APHA 2130 Part B.
1 Ea.