RPB™ T-Link™ Respirator with Bump Cap, Constant Flow
T-Link™ SAR system with bump cap and constant flow features a large visor, enhanced downward and peripheral vision, and fully adjustable design to promote safety and comfort.
Supplier:  RPB™ 1711112
Catalog No. 17000113
Explore available promotions
Alert:
WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Description
Description
  • Loose fitted respirator, no fit testing. Allows for facial hair and glasses without compromising safety.
  • Fixed lens frame ensures the lens will never hit your nose or optical aids being used.
  • APF 1000+
  • Air flow is fully adjustable with the air deflector system to suit personal preference and eliminate fogging.
  • All parts can be purchased individually for easy maintenance and servicing.
  • Large viewing window for exceptional downward and peripheral vision allowing for both verbal and non-verbal communication.
  • Optional tear-off lenses are available to protect visor lens and to quickly clear your field of view.
  • Soft, breathable inner-bib and long tuck-in collar for extra protection and upper body air flow.
  • Deconable head suspension system and disposable hood.
  • Padded interior with adjustable ratchet system ensures a perfect customized fit that supports user movement and increases comfort.
  • The optional Comfort-Link™ system provides extra comfort and stability, providing optimum weight distribution.
  • Breathing tube easily attaches to respirator and swivels to eliminate tangles. Centralized location ensures even weight distribution.
  • Two cape materials available: DuPont™ Tychem™ 2000 and DuPont™ Tychem™ 4000.
  • Tychem™ breathing tube cover available to increase the life and cleanability of the breathing tube.
  • Option of bump cap or ANSI Z89.1 hard hat protection (EN397).
Specifications
Specifications
ANSI S3.19, Z89.1 Type 1 Class C, Z87
CE2797, BSI Certified Product
White, Black
Supplied Air Respirator System
Yes
Tychem™ 2000 QC Hood (17-712), T-Link™ Bump Cap Assembly (17-110), Breathing Tube (04-830), Constant Flow Valve (NIOSH Approved) (03-101)
17-111-12
14 x 11 x 12 in.
1000
Yes
Tychem 2000, Plastic, Nylon, Nickel-plated Steel
One Size
Healthcare, Life Sciences, Painting, Coating, Pharma, Concrete Cutting, Aggregate, Pharmaceutical Manufacturing, Chemical, Pesticide, Materials Handling, Chemical Stripping, Sanding, Waste Management
PETG
14°F to 113°F (-10°C to +45°C), <90%RH
3.522 lb.