RPB™ T200™ Respirator with Full Hood with Internal Face Seal, Air Duct/Head Harness Assembly
T200™ SAR system equipped with a full hood, internal face seal, and air duct/head harness assembly supports true freedom of movement with its functional design, comfort, and durability.
Supplier:  RPB™ 1720032
Catalog No. 17000206
Explore available promotions
Alert:
WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Description
Description
  • Face Seal option allows access to ears for easy use with stethoscopes.
  • Moveable lens and air duct positioning to accommodate for facial features, optical aids and loupes.
  • APF 1000+
  • Multiple Tychem™ Hood styles available
  • Tychem™ breathing tube cover available to increase the life and cleanability of the breathing tube.
  • Large viewing window for exceptional downward and peripheral vision allowing for both verbal and non-verbal communication.
  • Optional tear-off lenses are available to protect visor lens.
  • Deconable head suspension system and disposable hood.
  • Breathing tube easily attaches to respirator and swivels to eliminate tangles. Centralized location ensures even weight distribution.
  • Customizable head suspension system includes an adjustable ratchet at the back and over-the-head straps to allow for a secure, individualized fit.
  • All parts can be purchased individually for easy maintenance and servicing.
Specifications
Specifications
ANSI S3.19, Z87
ISO9001
White
Supplied Air Respirator System
Healthcare, Life Sciences, Pharma
PETG
14°F to 113°F (-10°C to +45°C), <90%RH
2 lb.
1000
Yes
Tychem 2000, HDPE
One Size
Full Hood with Internal Face Seal, Air Duct/Head Harness Assembly (17-200-32)
17-200-32
14 x 10.5 x 6 in.