missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tacrolimus Monohydrate 80.0+%, TCI America™
$398.16 - $1381.95
Chemical Identifiers
CAS | 109581-93-3 |
---|---|
Molecular Formula | C44H71NO13 |
Molecular Weight (g/mol) | 822.046 |
MDL Number | MFCD11045918 |
InChI Key | NWJQLQGQZSIBAF-MLAUYUEBSA-N |
Synonym | FK 506 |
PubChem CID | 5282315 |
ChEBI | CHEBI:61057 |
SMILES | CC1CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC(=O)C(C=C(C1)C)CC=C)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC.O |
Chemical Identifiers
109581-93-3 | |
822.046 | |
NWJQLQGQZSIBAF-MLAUYUEBSA-N | |
5282315 | |
CC1CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC(=O)C(C=C(C1)C)CC=C)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC.O |
C44H71NO13 | |
MFCD11045918 | |
FK 506 | |
CHEBI:61057 |
Specifications
109581-93-3 | |
White | |
MFCD11045918 | |
2811 | |
NWJQLQGQZSIBAF-MLAUYUEBSA-N | |
822.046 | |
CHEBI:61057 | |
≥80.0% (HPLC) | |
Tacrolimus Monohydrate |
127°C | |
C44H71NO13 | |
10 mg | |
FK 506 | |
CC1CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC(=O)C(C=C(C1)C)CC=C)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC.O | |
5282315 | |
804.03 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : KD4201200
TSCA : Yes
Recommended Storage : Freezer