Sartorius Tacta™ Mechanical Pipette, Trade-In
Upgrade your pipetting with unbeatable prices. For a limited time, you can get up to 35% savings on Tacta Mechanical Pipettes.
Supplier:  Sartorius LH729050PR
 View more versions of this product
Catalog No. 14577796PR
Description
Description
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
Offer Details
After purchasing this trade-in program item please follow these steps to complete your trade in:
1. Complete Sartorius redemption form. (Contact Fisher Customer Service to obtain the form.)
2. Sartorius will email a link to a prepaid UPS shipping label, shipping instructions, and decontamination form.
3. Visit UPS site, print the shipping label, pack pipette, and ship to Sartorius.
Note: Any brand pipette, in any condition can be traded in. Pipettes must be free of contamination and returned together with the signed Decontamination Form.
Specifications
Specifications
Optifit Tips: 200, 350μL, SafetySpace Filter Tips: 120μL
Yellow
Tacta™
10 to 100 μL
Autoclavable
1
Pipette
Yellow