Sartorius Tacta™ Mechanical Pipettes
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
Supplier: Sartorius LH729010
Description
- Comfortable to hold due to the ergonomically designed handle
- Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
- Controlled and smooth tip ejection with the new Sartorius Optiject technology
- Calibration adjustment to provide accurate results for various liquid types
- Easy to clean, with only three parts to disassemble
- Reliable results, even over lengthy pipetting periods
Specifications
Optifit Tips: 10μL, SafetySpace Filter Tips: 10μL | |
Gray | |
Tacta™ | |
0.3 to 3 μL |
Autoclavable | |
1 | |
Pipette | |
Gray |