Sartorius Tacta™ Mechanical Pipettes
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
Supplier:  Sartorius LH729020
 View more versions of this product
Catalog No. 14-577-794
Explore available promotions
Description
Description
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
Specifications
Specifications
Optifit Tips: 10μL, SafetySpace Filter Tips: 10μL
Gray
Tacta™
1 to 10 μL
Autoclavable
1
Pipette
Gray
Videos
SDS
Documents
Documents
Accessories