Troemner™ Talboys™ 3-Prong Dual Adjust Nickel-Plated Zinc Clamp
Designed to securely hold every type of laboratory glassware and apparatus. The Troemner™ 3-Prong Dual Adjust Nickel-Plated Zinc Clamp is constructed with round extension arms, which allow the clamps to be rotated 360°.
Supplier: Troemner™ 30392198
Description
- Large grip adjustment range
- 3-prong construction
- Single or dual adjust
- Nickel-plated zinc
- Supplied with non-slip vinyl sleeves, and for temperatures above 100°C, fiberglass covers.
Specifications
3-Prong Dual Adjust Clamp | |
Nickel-plated Zinc |