missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Talc (Powder/USP), Fisher Chemical™

$321.38 - $321.38

Specifications

Chemical Name or Material Talc
Melting Point 800°C
CAS 14807-96-6,14808-60-7
Color White
Physical Form Powder/Solid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
T2-500
View Documents Promotion Details
Thermo Fisher Scientific
FLT2500
500 g Poly Bottle
Each for $321.38
Case of 6 Each for $1,928.28
 
Description

Description

Specifications

Specifications

Talc
14807-96-6,14808-60-7
Powder/Solid
MFCD00084820
USP
7.0% max. (USP)
800°C
White
3MgO4SiO2H2O
Soapstone, Talcum
Pass Test (USP)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'