missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tamoxifen, MP Biomedicals

Tamoxifen, White Powder

Supplier:  MP Biomedicals Inc 0215673891

 View more versions of this product

Catalog No. MP215673891


Explore available promotions

Description

Description

  • Tamoxifen has been used to facilitate the recombination of ect2flox allele in mouse organs
  • It has also been used to study its effect on lipopolysaccharide (LPS)-induced microglial activation

Specifications

Specifications

TAMOXIFEN
96°C to 100°C
C26H29NO
MFCD00010454
tamoxifen, trans-tamoxifen, crisafeno, soltamox, tamoxifene, diemon, tamoxifeno, tamoxifenum, citofen, istubol
NKANXQFJJICGDU-QPLCGJKRSA-N
2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
2733526
371.5
10540-29-1
White
C26H29NO
1 g
Practically insoluble in water (< 0.01% at 20°C); soluble in methanol, ethanol, 2-propanol, propylene glycol, chloroform (50mg/mL - clear, colorless to faint yellow solution) or DMSO.
CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
371.52
CHEBI:41774
Powder

Chemical Identifiers

10540-29-1
371.52
NKANXQFJJICGDU-QPLCGJKRSA-N
CHEBI:41774
C26H29NO
MFCD00010454
2733526
CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at +4°C.

SDS
Documents

Documents

Promotions

Promotions