missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tannic acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1702230
Description
Used for enzyme immobilization and protein adsorption.Tannic acid has anti-bacterial, anti-enzymatic and astringent applications. Used in the dyeing process for cellulose fibers such as cotton. Used in food and medication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed for enzyme immobilization and protein adsorption.Tannic acid has anti-bacterial, anti-enzymatic and astringent applications. Used in the dyeing process for cellulose fibers such as cotton. Used in food and medication.
Solubility
Soluble in water, warm glycerol, alcohol and acetone.
Notes
Air and light sensitive. Keep container tightly closed in a dry and well-ventilated place, cool place. Incompatible with metallic salts, strong oxidizing agents, iron, strong bases and heavy metals.
Specifications
Tannic acid | |
∼218°C (decomposition) | |
C76H52O46 | |
8186396 | |
tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6 | |
LRBQNJMCXXYXIU-JPZKJCTESA-N | |
1701.206 | |
1701.23 | |
Odorless |
3.5 | |
1401-55-4 | |
MFCD00066397 | |
14,9052 | |
Soluble in water,warm glycerol,alcohol and acetone. | |
C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C( | |
134129492 | |
250 g | |
Air and light sensitive |
Chemical Identifiers
1401-55-4 | |
1701.206 | |
LRBQNJMCXXYXIU-JPZKJCTESA-N | |
134129492 |
C76H52O46 | |
MFCD00066397 | |
tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6 | |
C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C( |
Safety and Handling
EINECSNumber : 215-753-2
RTECSNumber : WW5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only