missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tantalum ICP Standard, 1000 ppm Ta in H2O/tr HF, Ricca Chemical
Supplier: Ricca Chemical Company PTA1KW100
Specifications
Tantalum ICP Standard | |
Natural LDPE Bottle | |
98.0,0.19,0.1 | |
Colorless | |
F7H8N2Ta | |
RDNGDFQMYLJTJR-UHFFFAOYSA-I | |
[NH4+].[NH4+].F[Ta-2](F)(F)(F)(F)(F)F | |
350.015 |
12022-02-5,7664-39-3,7732-18-5 | |
100 mL | |
100.0,0.2,0.1 | |
1,000ppm Ta | |
1 mL = 1 mg Ta | |
Liquid | |
diazanium;heptafluorotantalum(2-) | |
71311477 |
Chemical Identifiers
12022-02-5 | |
350.015 | |
71311477 | |
[NH4+].[NH4+].F[Ta-2](F)(F)(F)(F)(F)F |
F7H8N2Ta | |
RDNGDFQMYLJTJR-UHFFFAOYSA-I | |
diazanium;heptafluorotantalum(2-) |