missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum powder, -325 mesh, 99.9% (metals basis), 99.6%, Thermo Scientific Chemicals

99.6% purity includes H, N, C, O

Supplier:  Thermo Scientific Chemicals 00033714

 View more versions of this product

Catalog No. AA0033714


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Black to Gray
7440-25-7
99.9% (Metals basis, 99.6% purity includes H, N, C, O)
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
CHEBI:33348
Odorless
Tantalum
Gray to black powder
Ta
25 g
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
-325 Mesh
Powder

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-135-5

RTECSNumber : WW5505000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only