Learn More
Tantalum(V) methoxide, Thermo Scientific Chemicals
$323.23 - $1,048.23
Chemical Identifiers
CAS | 865-35-0 |
---|---|
Molecular Formula | C5H15O5Ta |
Molecular Weight (g/mol) | 336.118 |
MDL Number | MFCD00058817 |
InChI Key | QASMZJKUEABJNR-UHFFFAOYSA-N |
PubChem CID | 12787641 |
IUPAC Name | methanolate;tantalum(5+) |
SMILES | C[O-].C[O-].C[O-].C[O-].C[O-].[Ta+5] |
Description
Tantalum(V) methoxide is used in the synthesis of tantalum hydride. Silica-supported alkoxo-tantalum complexes were used in the deperoxidation of cyclohexyl hydroperoxide into cyclohexanol and cyclohexanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
865-35-0 | |
336.118 | |
QASMZJKUEABJNR-UHFFFAOYSA-N | |
methanolate;tantalum(5+) |
C5H15O5Ta | |
MFCD00058817 | |
12787641 | |
C[O-].C[O-].C[O-].C[O-].C[O-].[Ta+5] |
Specifications
865-35-0 | |
189°C (10 mmHg) | |
Ta(OCH3)5 | |
5 g | |
Moisture sensitive | |
QASMZJKUEABJNR-UHFFFAOYSA-N | |
methanolate;tantalum(5+) | |
12787641 | |
Crystalline |
50°C | |
C5H15O5Ta | |
MFCD00058817 | |
UN3181 | |
Reacts with water. | |
C[O-].C[O-].C[O-].C[O-].C[O-].[Ta+5] | |
336.118 | |
336.12 | |
Tantalum(V) methoxide |
Safety and Handling
GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only