Applied Biosystems™ TaqCheck™ SARS-CoV-2 Control Dilution Buffer
Applied Biosystems TaqCheck SARS-CoV-2 Control Dilution Buffer is required for use of the TaqCheck SARS-CoV-2 Fast PCR Assay. It is used to dilute the RNA control.
Supplier: Applied Biosystems™ A50486
Specifications
1-step RT-qPCR | |
QuantStudio™ 5 RealâTime PCR Instrument, 384-well | |
3 x 250 μL | |
Human | |
Real Time PCR (qPCR) |
Fast | |
TaqCheck™ | |
Dry Ice | |
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) |
For Research Use Only. Not for use in diagnostic procedures.