missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan Fast Advanced Master Mix Gene Expression Bundle
TaqMan Fast Advanced Master Mix delivers better sensitivity, greater specificity, and faster results
$1279.28 - $1572.00
Specifications
| Description | Gene Expression Starter Kit |
|---|---|
| For Use With (Application) | Real Time PCR (qPCR) |
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
A47374
|
Applied Biosystems™
A47374 |
Master Mix |
Each for $1,572.00
|
|
|||||
A47377
|
Applied Biosystems™
A47377 |
Master Mix, MicroAmp Plates, and MicroAmp Seals |
Each for $1,327.64
|
|
|||||
A47376
|
Applied Biosystems™
A47376 |
Master Mix, MicroAmp Fast Plates, and MicroAmp Seals |
Each for $1,279.28
|
|
|||||
Description
Applied Biosystems has affordable qPCR tools that fit every budget and level of experience. Makes getting the best results from your data easy while saving money.
Specifications
| Gene Expression Starter Kit | |
| Real Time PCR (qPCR) |