missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Tartaric Acid, 10% (w/v), Ricca Chemical
Chemical Identifiers
CAS |
87-69-4 |
Molecular Formula |
C4H6O6 |
Molecular Weight (g/mol) |
150.09 |
MDL Number |
MFCD00064207 |
InChI Key |
FEWJPZIEWOKRBE-UHFFFAOYNA-N |
Synonym |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid |
PubChem CID |
444305 |
ChEBI |
CHEBI:15671 |
IUPAC Name |
2,3-dihydroxybutanedioic acid |
SMILES |
OC(C(O)C(O)=O)C(O)=O |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
87-69-4 |
|
150.09 |
|
FEWJPZIEWOKRBE-UHFFFAOYNA-N |
|
444305 |
|
2,3-dihydroxybutanedioic acid |
|
C4H6O6 |
|
MFCD00064207 |
|
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid |
|
CHEBI:15671 |
|
OC(C(O)C(O)=O)C(O)=O |
Specifications
|
0.89,0.0891 |
|
0.93,0.0928 |
|
C4H6O6 |
|
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid |
|
OC(C(O)C(O)=O)C(O)=O |
|
2,3-dihydroxybutanedioic acid |
|
444305 |
|
500 mL |
|
Liquid |
|
Colorless to Yellow |
|
7732-18-5 |
|
MFCD00064207 |
|
FEWJPZIEWOKRBE-UHFFFAOYNA-N |
|
10% (w/v) |
|
150.09 |
|
CHEBI:15671 |
|
Natural Poly Bottle |
|
Tartaric Acid |
SDS
missing translation for 'documents'