missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Tartrazine, 89%, pure
Supplier: Thermo Scientific Chemicals 191891000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tartrazine | |
>300°C | |
Orange | |
89% | |
C16H9N4Na3O9S2 | |
100 g | |
15, 9206 | |
Solubility in water: 260g/L water (30°C) | |
C1=CC(=CC=C1N=NC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)[O-])C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+] | |
534.35 | |
534.35 | |
Pure | |
Powder |
1934-21-0 | |
>300°C | |
Authentic | |
Glass bottle | |
MFCD00148908 | |
25, IV, 1757 | |
tartrazine, yellow 5, aizen tartrazine, trisodium salt, food yellow 4, tartrazine FD&C yellow #5, c.i. acid yellow 23, atul tartrazine, erio tartrazine, kako tartrazine | |
UJMBCXLDXJUMFB-UHFFFAOYSA-K | |
trisodium;5-oxo-1-(4-sulfonatophenyl)-4-[(4-sulfonatophenyl)diazenyl]-4H-pyrazole-3-carboxylate | |
164825 | |
89% | |
0.2% max. (in water) |
Chemical Identifiers
1934-21-0 | |
534.35 | |
UJMBCXLDXJUMFB-UHFFFAOYSA-K | |
164825 | |
C1=CC(=CC=C1N=NC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)[O-])C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+] |
C16H9N4Na3O9S2 | |
MFCD00148908 | |
tartrazine, yellow 5, aizen tartrazine, trisodium salt, food yellow 4, tartrazine FD&C yellow #5, c.i. acid yellow 23, atul tartrazine, erio tartrazine, kako tartrazine | |
trisodium;5-oxo-1-(4-sulfonatophenyl)-4-[(4-sulfonatophenyl)diazenyl]-4H-pyrazole-3-carboxylate |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 217-699-5
RUO – Research Use Only