missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lovibond™ TB350 IR Turbidimeter Kit

Ideal for field and environmental testing, the TB350 IR portable turbidimeter delivers the most reliable measurements for low range to high range samples without sacrificing accuracy.

Supplier:  Lovibond™ 194300

Catalog No. 09054112


Explore available promotions

Description

Description

  • Reads up to 4000 NTU without the need for dilutions
  • Superior data management - easily save and recall measurement data sets
  • Touchscreen interface for ease of operation
  • ISO 7027 Compliant
  • Supplied with AA
  • Rechargeable battery pack accessory available

The Lovibond™ TB Series of turbidimeters offers simplistic operation combined with intelligent instrument engineering to provide an unparalleled level of accuracy in turbidity measurement.

Featuring the patent-pending Multipath 90° BLAC™ sensor technology, the optical system is engineered with dual detectors to deliver a ratio reading which mitigates common measurement stability issues.

The intuitive, touchscreen interface makes it easy to perform procedures and interpret results. With function applications, you can navigate between settings and performing that function efficiently. This user interface helps eliminates common frustrations and prevents errors. The data logging capabilities allow you to record the testing location, operator's identification, time and date along with the measurement. Stored data can be transferred to a computer via USB.

Specifications

Specifications

Portable Turbidity Meter
Color Touch Screen
Yes
Instrument, calibration standards, verification standard, silicone oil, 2x sample cells with quick start guide and Certificate of Conformity in a carrying case.
ABS
User Selectable
IP67
±1.8% plus Stray Light
0 to 4000 NTU/FNU
NTU, NFU
500 Tests
ISO 7027
1 Ea.
Infrared LED
7 sec.
Zero plus 3 points
Measurement, Calibration, Verification and Logging
10 mL
Portable and/or field applications when expected turbidity is between 0 and 4000 NTU.
Included with kit purchase. Replacement sets available.
Better than 1% or ±0.01 NTU
0.01
17 x 15 x 5.5 in.
AA or Rechargeable Battery Pack
7027
Videos
SDS
Documents

Documents

Promotions

Promotions