missing translation for 'onlineSavingsMsg'
Learn More

TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™

CAS: 10043-35-3

Supplier:  Fisher BioReagents V36401

 View more versions of this product

Catalog No. BP1333-1

Only null left
Explore available promotions

Description

Description

TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TBE buffer solution - simply dilute as needed
  • 10X solution contains 0.89M tris, 0.89M borate, and 0.02M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Functionally tested for use in electrophoresis applications
  • TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
  • Intended for laboratory research use only
Specifications

Specifications

Tris-Borate-EDTA
10043-35-3,60-00-4,77-86-1,7732-18-5
8.2 to 8.4
1 L
Filtered through a 0.2-micron filter.
Nalgene Poly Bottle
TBE
10X Solution
Colorless
Liquid
DNase free
DNase-, RNase- and Protease-Free
Protease free
Electrophoresis

Chemical Identifiers

10043-35-3
TBE
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions