missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TBS, Tris Buffered Saline, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
$186.58 - $186.58
Specifications
Chemical Name or Material | Tris Buffered Saline |
---|---|
Name Note | 1X Solution, pH 7.4 |
CAS | 7647-14-5,7447-40-7,7647-01-0,7732-18-5,77-86-1 |
Boiling Point | 100°C |
Color | Colorless |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP24721
|
Fisher BioReagents
BP24721 |
1 L | Nalgene Poly Bottle |
Each for $186.58
Case of 6 Each for $1,119.48
|
|
||||
Description
Tris Buffered Saline (TBS) is used in biochemical and molecular biology applications.Specifications
Tris Buffered Saline | |
7647-14-5,7447-40-7,7647-01-0,7732-18-5,77-86-1 | |
Colorless | |
DNase free | |
TBS |
1X Solution, pH 7.4 | |
100°C | |
Liquid | |
Protease free | |
Molecular Biology |