missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TCLP Extraction Fluid No. 2, Certified, LabChem™
Supplier: LabChem LC259974
Specifications
TCLP Extraction Fluid No. 2 | |
Amber Glass | |
99.43,0.57 | |
CH3COOH | |
Certified | |
1.02cSt | |
2.88 ±0.05 | |
Liquid |
64-19-7,7732-18-5 | |
4 L | |
Colorless | |
60.05 | |
Passes Test | |
0.999g/mL | |
Soluble in water |