missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tebbe reagent, 0.5M solution in toluene, Thermo Scientific Chemicals
$422.51 - $422.51
Specifications
Chemical Name or Material | Tebbe reagent |
---|---|
Name Note | 0.5M solution in toluene |
CAS | 67719-69-1,108-88-3 |
Color | Purple to Red |
Density | 0.9270g/mL |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC368970250
|
Thermo Scientific Chemicals
368970250 |
25 mL | Glass Bottle |
Each for $422.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination), Lewis acidSpecifications
Tebbe reagent | |
67719-69-1,108-88-3 | |
0.9270g/mL | |
C13H18AlClTi | |
08,83; 10,87; 11,52; 12,54; 13,71; 15,79; 16,70; 17,69 | |
Bis(cyclopentadienyl-μ-chloro(dimethylaluminum-μ-methylenetitanium | |
284.62 |
0.5M solution in toluene | |
Purple to Red | |
4°C | |
MFCD00151575 | |
0.927 | |
Solubility in water: reacts. | |
Solution |
RUO – Research Use Only