missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tecniplast™ Deluxe Reusable Animal Cages

Safe, sanitary environments for mice, rats, hamsters, guinea pigs and gerbils

Supplier:  Tecniplast™ 1290D001

 View more versions of this product

Catalog No. 01-288-1D


Explore available promotions

Description

Description

  • Sturdy cages feature rounded corners for easy cleaning
  • Corner lugs or side arms allow stacking without scratching or jamming
  • Filter covers (sold separately) maintain environment for optimum animal health; lock in place to assure containment Durable wire lids (sold separately) available for several cage sizes
  • Tecniplast water bottles (sold separately) available for most cage sizes; fit securely beneath filter cove

Reusable Polycarbonate Cages

  • Withstand temperatures up to 121°C (250°F)
  • Glass-like clarity with excellent impact strength
  • Autoclavable
Reusable High-Temperature Polysulfone Cages
  • Withstand repeated autoclaving at 134°C (273°F)
  • Molded of clear, special-grade PSF to withstand higher temperatures and provide better chemical resistance

Order Info

Lids and accessories sold separately.

Compliance

Meet NIH size guidelines

Specifications

Specifications

Polycarbonate
250°F
16.75L x 10.5W x 6 in. H (42.5 x 26.6 x 15.24 cm)
SDS
Documents

Documents

Promotions

Promotions