missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tecniplast™ Stand for Metabolic Cages

Stainless-steel cradle assembly with nylon supports

Supplier:  Tecniplast™ 3M01D100

Catalog No. 01-287-6E


Explore available promotions

Description

Description

  • Autoclavable

Order Info

Overall dimensions: 12.25L x 12.25W x 13.37 in.H (31 x 31 x 34cm).

Specifications

Specifications

Cage Stand
Metabolic cage
12.2 in.
12.2 in.
13.39 in.
Stainless Steel
12.2 x 12.2 x 13.39 in. (3.1 x 31 x 3.4 cm)
3.1 cm
31 cm
3.4 cm
Videos
SDS
Documents

Documents

Promotions

Promotions