MilliporeSigma™ Supelco™ Tedlar™ SCV Gas Sampling Bag with Thermogreen™ LB-2 Septa
Tedlar™ is recommended in several US EPA methods for sampling VOCs and sulfur compounds.
Supplier: MilliporeSigma™ Supelco™ 30284U
Description
Tedlar™ is made from a high-quality 2 mil Tedlar™ film for superior inertness, impermeability, sample integrity and good recovery data.Specifications
0.6 L | |
PVDF Film | |
10/Pk. |
Screw Cap Valve (SCV) | |
Tedlar |
Tedlar is a registered trademark of E. I. du Pont de Nemours and Company. Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC.