missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Teflon Coated Spatula

Features one flat end and one spoon end with Teflon coating that provides a chemical resistant, non-stick surface

Supplier:  Eisco™ CH0635TFC

Catalog No. S27088


Explore available promotions

Description

Description

  • Teflon Coated Spatula
  • includes one flat end and one spoon end
  • Chemical resistant
  • Non-stick
  • Overall length: 9 in.
  • Perfect for any science or home laboratory
Specifications

Specifications

1
Teflon
9 in.
Stainless Steel
Autoclavable
Double Ended Spatula
9 mm (Spatula), 15 mm (Spoon)
23 cm
Videos
SDS
Documents

Documents

Promotions

Promotions