Learn More
Thermo Scientific Chemicals Teicoplanin, cont. 80+% A2
CAS: 61036-62-21879.67 g/mol
$229.49 - $716.71
Chemical Identifiers
CAS 61036-62-2
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66785MC
View Documents
Thermo Scientific Chemicals
J66785MC
100mg
Each for $229.49
Only null left
AAJ66785ME
View Documents
Thermo Scientific Chemicals
J66785ME
500mg
Each for $716.71
Only null left
Description
Description
A glycopeptide antibiotic produced by Actinoplanes teichomiceticus; effective against Gram-positive bacteria that are resistant to β-lactam antibiotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Identifiers
61036-62-2
Specifications
80%
Keep cold
1879.67
100mg
61036-62-2
Powder
Teicoplanin
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
RUO – Research Use Only