missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tellurium ingot/button,≈38mm (1.5 in.) dia. x 6mm (0.24 in.) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045108KF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver | |
≈38mm (1.5 in.) dia. x 6mm (0.24 in.) thick | |
(metals basis) | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
[Te] | |
127.60 | |
CHEBI:30452 | |
Odorless | |
Pieces |
Tellurium ingot/button | |
13494-80-9 | |
Te | |
1 Pc. | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
tellurium | |
6327182 | |
99.9% | |
≈30g/piece |
Chemical Identifiers
13494-80-9 | |
127.60 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
6327182 | |
tellurium |
Te | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
CHEBI:30452 | |
[Te] |
Safety and Handling
GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
P264b-P270-P301+P310-P330-P501c
H301
EINECSNumber : 236-813-4
RTECSNumber : WY2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only