Learn More
Tellurium(IV) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
$225.92 - $688.47
Chemical Identifiers
CAS | 10026-07-0 |
---|---|
Molecular Formula | Cl4Te |
Molecular Weight (g/mol) | 269.4 |
MDL Number | MFCD00011262 |
InChI Key | SWLJJEFSPJCUBD-UHFFFAOYSA-N |
Synonym | tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4 |
PubChem CID | 61443 |
SMILES | Cl[Te](Cl)(Cl)Cl |
Description
Tellurium(IV) chloride is used in organic synthesis to prepare Cl-C-C-TeCl3derivatives by reacting with alkenes and aryl tellurium compounds by reacting with electron rich arenes. It is involved in the preparation of diaryl telluride from anisole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10026-07-0 | |
269.4 | |
SWLJJEFSPJCUBD-UHFFFAOYSA-N | |
61443 |
Cl4Te | |
MFCD00011262 | |
tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4 | |
Cl[Te](Cl)(Cl)Cl |
Specifications
380°C | |
10026-07-0 | |
25 g | |
Cl4Te | |
tellurium tetrachloride, telluric chloride, tellurium chloride, tetrachlorotellurium, tellurium iv chloride, tellurium chloride tecl4 , t-4, unii-dny2r5498h, tellurium chloride, t-4, telluricchloride, tecl4 | |
SWLJJEFSPJCUBD-UHFFFAOYSA-N | |
269.4 | |
269.41 | |
3.01 g/mL |
224°C | |
Crystalline | |
99.9% (metals basis) | |
MFCD00011262 | |
Decomposes in water forming TeO2 and HClSoluble in acetonitrile,dimethyl sulfoxide and dimethylformamide. Slightly soluble in ether and benzene. Insoluble in cyclohexane,hexanes,methylene chloride and chloroform. | |
Cl[Te](Cl)(Cl)Cl | |
61443 | |
Moisture sensitive | |
Tellurium(IV) chloride |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 233-055-6
RTECSNumber : WY2635000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only