Learn More
Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals
$911.07 - $911.07
Chemical Identifiers
CAS | 7446-07-3 |
---|---|
Molecular Formula | O2Se |
Molecular Weight (g/mol) | 110.97 |
MDL Number | MFCD00003562 |
InChI Key | JPJALAQPGMAKDF-UHFFFAOYSA-N |
Synonym | selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4 |
PubChem CID | 24007 |
IUPAC Name | (oxo-λâ´-selanylidene)oxidane |
SMILES | O=[Se]=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC222592500
|
Thermo Scientific Chemicals
222592500 |
250 g | Glass bottle |
Each for $911.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-07-3 | |
110.97 | |
JPJALAQPGMAKDF-UHFFFAOYSA-N | |
24007 | |
O=[Se]=O |
O2Se | |
MFCD00003562 | |
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4 | |
(oxo-λâ´-selanylidene)oxidane |
Specifications
1245°C | |
7446-07-3 | |
Powder | |
99+% | |
TeO2 | |
15, 9265 | |
Solubility in water: insoluble. Other solubilities: soluble in conc. acids and alkalies,slightly soluble in diluted acids | |
O=[Se]=O | |
110.97 | |
159.6 | |
Glass bottle | |
Tellurium(IV) oxide, 99+% |
733°C | |
White to Yellow | |
250 g | |
O2Se | |
MFCD00003562 | |
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4 | |
JPJALAQPGMAKDF-UHFFFAOYSA-N | |
(oxo-λâ´-selanylidene)oxidane | |
24007 | |
≥99% | |
5.67g/mL |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if inhaled.
May damage fertility or the unborn child.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio
GHS Signal Word: Danger
EINECSNumber : 231-193-1
RTECSNumber : WY2675000
TSCA : TSCA
RUO – Research Use Only