missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tellurium pieces, 99.9999% (metals basis), zone refined, Thermo Scientific Chemicals
$133.79 - $1930.25
Chemical Identifiers
CAS | 13494-80-9 |
---|---|
Molecular Formula | Te |
Molecular Weight (g/mol) | 127.60 |
MDL Number | MFCD00134062 |
InChI Key | PORWMNRCUJJQNO-UHFFFAOYSA-N |
Synonym | tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds |
PubChem CID | 6327182 |
ChEBI | CHEBI:30452 |
IUPAC Name | tellurium |
SMILES | [Te] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1107609
|
Thermo Scientific Chemicals
01107609 |
10 g |
Each for $133.79
|
|
|||||
AA1107618
|
Thermo Scientific Chemicals
01107618 |
50 g |
Each for $481.60
|
|
|||||
AA1107630
|
Thermo Scientific Chemicals
01107630 |
250 g |
Each for $1,930.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13494-80-9 | |
127.60 | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
6327182 | |
tellurium |
Te | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
CHEBI:30452 | |
[Te] |
Specifications
13494-80-9 | |
MFCD00134062 | |
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds | |
[Te] | |
127.60 | |
CHEBI:30452 | |
Odorless | |
Tellurium pieces, Zone refined |
Te | |
10 g | |
PORWMNRCUJJQNO-UHFFFAOYSA-N | |
tellurium | |
6327182 | |
99.9999% | |
Pieces | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
P264b-P270-P301+P310-P330-P501c
H301
EINECSNumber : 236-813-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only